siRNA Details
| Gene Symbol: PPP2R3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190447.1 | NP_001177376.1 | 13 | 2468 | ||
| NM_002718.4 | NP_002709.2 | 14 | 4903 | |||
| NM_181897.2 | NP_871626.1 | 13 | 2758 | |||
| XM_006713686.3 | XP_006713749.1 | 14 | 5314 | |||
| XM_011512956.2 | XP_011511258.1 | 14 | 5078 | |||
| XM_017006787.1 | XP_016862276.1 | 13 | 4636 | |||
| GenBank mRNA | AL389975.1 | - | 11 | 1986 | ||
| L07590.1 | - | 14 | 4788 | |||

PPP2R3A
Genomic Map