siRNA Details
| Gene Symbol: PTPN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145368.1 | NP_001138840.1 | 25 | 3755 | ||
| NM_001145369.1 | NP_001138841.1 | 21 | 5939 | |||
| NM_001145370.1 | NP_001138842.1 | 20 | 5804 | |||
| NM_001145371.1 | NP_001138843.1 | 16 | 3373 | |||
| NM_001145372.1 | NP_001138844.1 | 15 | 3238 | |||
| NM_002829.3 | NP_002820.3 | 26 | 3890 | |||
| NR_026918.1 | 13 | 2660 | ||||
| XM_006717197.3 | XP_006717260.1 | 26 | 3999 | |||
| XM_006717199.3 | XP_006717262.1 | 26 | 4024 | |||
| XM_006717201.3 | XP_006717264.1 | 25 | 3831 | |||
| XM_006717202.3 | XP_006717265.1 | 23 | 3593 | |||
| XM_006717203.3 | XP_006717266.1 | 23 | 3552 | |||
| XM_006717204.3 | XP_006717267.1 | 26 | 3926 | |||
| XM_011518888.2 | XP_011517190.1 | 26 | 3960 | |||
| XM_011518889.2 | XP_011517191.1 | 26 | 4630 | |||
| XM_017014955.1 | XP_016870444.1 | 26 | 3982 | |||
| XM_017014956.1 | XP_016870445.1 | 24 | 3703 | |||
| XM_017014957.1 | XP_016870446.1 | 25 | 3791 | |||
| GenBank mRNA | BC033716.1 | - | 8 | 1916 | ||
| BX648253.1 | - | 20 | 5805 | |||
| BX648735.1 | - | 21 | 4374 | |||
| CR749204.1 | - | 14 | 2817 | |||
| M64572.1 | - | 25 | 3798 | |||

PTPN3
Genomic Map