siRNA Details
| Gene Symbol: PTPRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161440.2 | NP_001154912.2 | 6 | 1207 | ||
| NM_002842.4 | NP_002833.4 | 8 | 1741 | |||
| XM_011527183.1 | XP_011525485.1 | 8 | 1762 | |||
| XM_011527188.1 | XP_011525490.1 | 7 | 1711 | |||
| XM_011527190.1 | XP_011525492.1 | 8 | 1762 | |||
| XM_017027056.1 | XP_016882545.1 | 7 | 1749 | |||
| XM_017027057.1 | XP_016882546.1 | 7 | 1749 | |||
| XM_017027058.1 | XP_016882547.1 | 7 | 1782 | |||
| XM_017027059.1 | XP_016882548.1 | 7 | 1749 | |||
| XM_017027060.1 | XP_016882549.1 | 7 | 1749 | |||
| XM_017027061.1 | XP_016882550.1 | 5 | 1215 | |||
| XM_017027062.1 | XP_016882551.1 | 7 | 1657 | |||
| XM_017027063.1 | XP_016882552.1 | 7 | 1650 | |||
| XM_017027064.1 | XP_016882553.1 | 7 | 1749 | |||
| GenBank mRNA | AK097257.1 | - | 8 | 1760 | ||
| BC111715.1 | - | 7 | 1157 | 40008777 | ||
| BC111716.1 | - | 8 | 1691 | 40008775 | ||
| D15049.2 | - | 8 | 1715 | |||

PTPRH
Genomic Map