siRNA Details
| Gene Symbol: TRPM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177310.1 | NP_001170781.1 | 13 | 1515 | ||
| NM_001177311.1 | NP_001170782.1 | 13 | 1542 | |||
| NM_017662.4 | NP_060132.3 | 13 | 1696 | |||
| XM_011518244.1 | XP_011516546.1 | 13 | 1696 | |||
| XM_011518245.1 | XP_011516547.1 | 13 | 1696 | |||
| XM_011518246.1 | XP_011516548.1 | 13 | 1696 | |||
| XM_011518247.1 | XP_011516549.1 | 13 | 1696 | |||
| XM_011518248.1 | XP_011516550.1 | 13 | 1696 | |||
| XM_011518249.1 | XP_011516551.1 | 13 | 1696 | |||
| XM_011518250.1 | XP_011516552.1 | 13 | 1696 | |||
| XM_011518251.2 | XP_011516553.1 | 8 | 947 | |||
| XM_011518252.2 | XP_011516554.1 | 13 | 1696 | |||
| XM_011518255.2 | XP_011516557.1 | 13 | 1696 | |||
| XM_017014287.1 | XP_016869776.1 | 13 | 1696 | |||
| XM_017014288.1 | XP_016869777.1 | 13 | 1696 | |||
| XM_017014289.1 | XP_016869778.1 | 13 | 1696 | |||
| GenBank mRNA | AF350881.2 | - | 13 | 1696 | ||
| AF448232.1 | - | 13 | 1458 | |||
| AK026281.1 | - | 7 | 763 | |||
| AY333282.1 | - | 13 | 1521 | |||
| AY333283.1 | - | 13 | 1515 | |||
| AY333284.1 | - | 13 | 1542 | |||
| AY333285.1 | - | 13 | 1521 | |||
| AY333286.1 | - | 13 | 1521 | |||
| AY333287.1 | - | 13 | 1521 | |||

TRPM6
Genomic Map