siRNA Details
| Gene Symbol: MTMR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004685.3 | NP_004676.3 | 13 | 1814 | ||
| XM_011535306.1 | XP_011533608.1 | 14 | 1874 | |||
| XM_017020846.1 | XP_016876335.1 | 10 | 1079 | |||
| GenBank mRNA | AK025507.1 | - | 3 | 204 | ||

MTMR6
Genomic Map