siRNA Details
| Gene Symbol: CACHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293274.1 | NP_001280203.1 | 7 | 1292 | ||
| NM_020925.2 | NP_065976.2 | 8 | 1013 | |||
| XM_011541862.1 | XP_011540164.1 | 7 | 1288 | |||
| XM_011541863.1 | XP_011540165.1 | 8 | 1437 | |||
| GenBank mRNA | AB046793.1 | - | 6 | 795 | ||
| BC039301.1 | - | 8 | 999 | 4836806 | ||
| BX537603.1 | - | 7 | 1291 | |||

CACHD1
Genomic Map