siRNA Details
| Gene Symbol: PPP2R5E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282179.1 | NP_001269108.1 | 14 | 2681 | ||
| NM_001282180.1 | NP_001269109.1 | 14 | 2669 | |||
| NM_001282181.1 | NP_001269110.1 | 13 | 2051 | |||
| NM_001282182.1 | NP_001269111.1 | 13 | 2006 | |||
| NM_006246.3 | NP_006237.1 | 14 | 2684 | |||
| NR_104104.1 | 13 | 2487 | ||||
| GenBank mRNA | BC034994.1 | - | 15 | 2678 | ||
| BQ439131.1 | - | 1 | 178 | |||
| L76703.1 | - | 14 | 2662 | |||
| Z69029.1 | - | 14 | 2597 | |||

PPP2R5E
Genomic Map