siRNA Details
| Gene Symbol: CLK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020666.2 | NP_065717.1 | 9, 10 | 1157 | ||
| GenBank mRNA | AF294429.1 | - | 8, 9 | 1049 | ||
| AK311485.1 | - | 8, 9 | 1068 | |||
| BC015942.1 | - | 9, 10 | 1157 | |||
| BC136261.1 | - | 9, 10 | 1105 | 9020248 | ||
| BC151233.1 | - | 9, 10 | 2371 | |||
| CR749504.1 | - | 11, 12 | 2036 | |||

CLK4
Genomic Map