siRNA Details
| Gene Symbol: PLPPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170635.1 | NP_001164106.1 | 10 | 1971 | ||
| NM_022737.2 | NP_073574.2 | 10 | 2030 | |||
| XM_005260017.1 | XP_005260074.1 | 10 | 2046 | |||
| XM_017027147.1 | XP_016882636.1 | 10 | 2049 | |||
| XM_017027148.1 | XP_016882637.1 | 10 | 1967 | |||
| GenBank mRNA | AK023117.1 | - | 10 | 1991 | ||
| AK094869.1 | - | 10 | 1859 | |||
| AK225482.1 | - | 9 | 1468 | |||
| AL353953.1 | - | 2 | 669 | |||
| AY304516.1 | - | 10 | 1985 | |||
| AY339628.1 | - | 9 | 1685 | |||
| BC009378.1 | - | 10 | 1873 | 4125316 | ||
| CR936652.1 | - | 5 | 2436 | |||

PLPPR2
Genomic Map