siRNA Details
| Gene Symbol: SCYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048218.1 | NP_001041683.1 | 7 | 1003 | ||
| NM_020680.3 | NP_065731.3 | 7 | 1003 | |||
| XM_005274118.3 | XP_005274175.1 | 7 | 1015 | |||
| XM_005274120.3 | XP_005274177.1 | 7 | 1015 | |||
| XM_005274121.3 | XP_005274178.1 | 7 | 1015 | |||
| XM_017018029.1 | XP_016873518.1 | 7 | 1015 | |||
| XM_017018030.1 | XP_016873519.1 | 7 | 1015 | |||
| XM_017018031.1 | XP_016873520.1 | 7 | 1015 | |||
| XM_017018032.1 | XP_016873521.1 | 7 | 857 | |||
| XM_017018033.1 | XP_016873522.1 | 7 | 1007 | |||
| GenBank mRNA | AB047077.1 | - | 7 | 926 | ||
| AB051427.1 | - | 7 | 926 | |||
| AB051428.1 | - | 7 | 926 | |||
| AF297709.1 | - | 7 | 965 | |||
| AK314945.1 | - | 7 | 959 | |||
| BC069233.1 | - | 7 | 967 | 6581110 | ||

SCYL1
Genomic Map