siRNA Details
| Gene Symbol: CD69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001781.2 | NP_001772.1 | 3 | 370 | ||
| GenBank mRNA | AK291869.1 | - | 3 | 370 | ||
| AK303174.1 | - | 3 | 372 | |||
| AK303383.1 | - | 3 | 370 | |||
| AY238518.1 | - | 3 | 289 | |||
| BC007037.1 | - | 3 | 370 | 3950403 | ||
| L07555.1 | - | 3 | 362 | |||
| Z22576.1 | - | 3 | 370 | |||

CD69
Genomic Map