siRNA Details
| Gene Symbol: CD72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001782.2 | NP_001773.1 | 8 | 1113 | ||
| XM_017015351.1 | XP_016870840.1 | 9 | 1486 | |||
| GenBank mRNA | AB209587.1 | - | 6 | 2905 | ||
| AF283777.2 | - | 7 | 1372 | |||
| BC011734.2 | - | 8 | 1024 | |||
| BC030227.1 | - | 8 | 1054 | 5226648 | ||
| DQ174771.1 | - | 9 | 1482 | |||
| M54992.1 | - | 8 | 1097 | |||

CD72
Genomic Map