siRNA Details
| Gene Symbol: CD27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242.4 | NP_001233.1 | 3 | 564 | ||
| XM_011521042.2 | XP_011519344.1 | 2 | 728 | |||
| XM_017020232.1 | XP_016875721.1 | 3 | 582 | |||
| XM_017020233.1 | XP_016875722.1 | 2 | 740 | |||
| XM_017020234.1 | XP_016875723.1 | 3 | 582 | |||
| GenBank mRNA | AK315732.1 | - | 3 | 583 | ||
| BC012160.1 | - | 3 | 550 | 4575359 | ||
| BI225100.1 | - | 3 | 610 | |||
| M63928.1 | - | 3 | 453 | |||

CD27
Genomic Map