siRNA Details
| Gene Symbol: CTSV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201575.1 | NP_001188504.1 | 8 | 1169 | ||
| NM_001333.3 | NP_001324.2 | 8 | 1372 | |||
| GenBank mRNA | AB001928.1 | - | 8 | 1195 | ||
| AF070448.1 | - | 7 | 1149 | |||
| AY358641.1 | - | 8 | 1372 | |||
| BC023504.2 | - | 8 | 1255 | 3139314 | ||
| Y14734.1 | - | 8 | 1191 | |||

CTSV
Genomic Map