siRNA Details
| Gene Symbol: CTSE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317331.1 | NP_001304260.1 | 7 | 1836 | ||
| NM_001910.3 | NP_001901.1 | 9 | 2135 | |||
| NM_148964.2 | NP_683865.1 | 8 | 1993 | |||
| XM_011509244.1 | XP_011507546.1 | 9 | 2140 | |||
| XM_011509245.1 | XP_011507547.1 | 8 | 1998 | |||
| GenBank mRNA | AJ250716.1 | - | 8 | 1979 | ||
| AJ250717.1 | - | 9 | 2121 | |||
| BC042537.1 | - | 9 | 2102 | 5174814 | ||
| J05036.1 | - | 9 | 2066 | |||

CTSE
Genomic Map