siRNA Details
| Gene Symbol: PSMB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002801.3 | NP_002792.1 | 8 | 964 | ||
| GenBank mRNA | AK225658.1 | - | 7 | 751 | ||
| BC017198.1 | - | 8 | 915 | 3143189 | ||
| BC052369.1 | - | 8 | 902 | 4054165 | ||
| Y13640.1 | - | 8 | 924 | |||

PSMB10
Genomic Map