siRNA Details
| Gene Symbol: USP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017415.1 | NP_001017415.1 | 5, 6 | 893 | ||
| NM_001017416.1 | NP_001017416.1 | 5, 6 | 798 | |||
| NM_003368.4 | NP_003359.3 | 5, 6 | 1323 | |||
| GenBank mRNA | AB014458.1 | - | 5, 6 | 674 | ||
| AB208893.1 | - | 5, 6 | 955 | |||
| AF117386.1 | - | 5, 6 | 810 | |||
| AK312423.1 | - | 5, 6 | 986 | |||
| AL117575.1 | - | 4, 5 | 645 | |||
| BC018745.1 | - | 5, 6 | 1323 | |||
| BC023216.1 | - | 1, 2 | 158 | |||
| BC032364.1 | - | 5, 6 | 973 | |||
| BC050525.1 | - | 5, 6 | 1027 | 6473568 | ||

USP1
Genomic Map