siRNA Details
| Gene Symbol: USP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017415.1 | NP_001017415.1 | 9 | 2503 | ||
| NM_001017416.1 | NP_001017416.1 | 9 | 2408 | |||
| NM_003368.4 | NP_003359.3 | 9 | 2933 | |||
| GenBank mRNA | AB014458.1 | - | 9 | 2284 | ||
| AB208893.1 | - | 9 | 2564 | |||
| AF117386.1 | - | 9 | 2420 | |||
| AK096788.1 | - | 1 | 1215 | |||
| AK312423.1 | - | 9 | 2596 | |||
| AL117503.1 | - | 1 | 472 | |||
| AL117575.1 | - | 8 | 2255 | |||
| BC023216.1 | - | 5 | 1768 | |||
| BC050525.1 | - | 9 | 2637 | 6473568 | ||

USP1
Genomic Map