siRNA Details
| Gene Symbol: USP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004651.3 | NP_004642.2 | 4 | 581 | ||
| XM_005272674.3 | XP_005272731.1 | 4 | 418 | |||
| XM_011543988.1 | XP_011542290.1 | 4 | 375 | |||
| GenBank mRNA | AB073597.1 | - | 4 | 483 | ||
| AK055588.1 | - | 3 | 638 | |||
| AK098394.1 | - | 4 | 485 | |||
| AK294635.1 | - | 4 | 327 | |||
| BC000350.2 | - | 4 | 455 | |||
| BC063668.1 | - | 4 | 463 | |||
| BC140849.1 | - | 4 | 630 | 9021717 | ||
| U44839.1 | - | 4 | 441 | |||

USP11
Genomic Map