siRNA Details
Gene Symbol: USP33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015017.4 | NP_055832.3 | 9 | 1034 | ||
NM_201624.2 | NP_963918.1 | 8 | 855 | |||
NM_201626.2 | NP_963920.1 | 9 | 1034 | |||
XM_005270648.1 | XP_005270705.1 | 8 | 682 | |||
XM_005270649.1 | XP_005270706.1 | 8 | 682 | |||
XM_011541055.1 | XP_011539357.1 | 8 | 682 | |||
XM_011541056.2 | XP_011539358.1 | 8 | 682 | |||
XM_017000722.1 | XP_016856211.1 | 8 | 855 | |||
XM_017000723.1 | XP_016856212.1 | 8 | 806 | |||
XM_017000724.1 | XP_016856213.1 | 8 | 855 | |||
XM_017000725.1 | XP_016856214.1 | 8 | 855 | |||
XM_017000726.1 | XP_016856215.1 | 8 | 855 | |||
XM_017000727.1 | XP_016856216.1 | 9 | 750 | |||
XM_017000728.1 | XP_016856217.1 | 9 | 712 | |||
XM_017000729.1 | XP_016856218.1 | 8 | 682 | |||
GenBank mRNA | AB029020.1 | - | 8 | 776 | ||
AF383172.1 | - | 9 | 1007 | |||
AF383173.1 | - | 8 | 828 | |||
AK022864.1 | - | 9 | 1007 | |||
AK225499.1 | - | 6 | 475 | |||
AK308402.1 | - | 8 | 746 | |||
BC016663.1 | - | 9 | 888 | 3874822 |

More Information
Matching Taqman Gene Expression Assay:
Panther Classification:
Gene Ontology Categories:
