siRNA Details
| Gene Symbol: USP47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282659.1 | NP_001269588.1 | 8 | 1629 | ||
| NM_017944.3 | NP_060414.3 | 7 | 1425 | |||
| XM_005252997.1 | XP_005253054.1 | 9 | 1689 | |||
| XM_017017953.1 | XP_016873442.1 | 9 | 1745 | |||
| GenBank mRNA | AK027283.1 | - | 537 | |||
| AK027362.1 | - | 7 | 830 | |||
| AK092290.1 | - | 9 | 1046 | |||
| AK127264.1 | - | 7 | 1425 | |||
| AK309240.1 | - | 8 | 1093 | |||

USP47
Genomic Map