siRNA Details
| Gene Symbol: CHN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025201.3 | NP_001020372.2 | 6 | 588 | ||
| NM_001822.5 | NP_001813.1 | 5 | 587 | |||
| NR_038133.1 | 5 | 401 | ||||
| GenBank mRNA | AB209788.1 | - | 5 | 386 | ||
| AK289941.1 | - | 5 | 517 | |||
| AK300890.1 | - | 6 | 582 | |||
| BC011393.1 | - | 5 | 559 | 3871858 | ||
| BQ893027.1 | - | 6 | 621 | |||
| Z22641.1 | - | 5 | 256 | |||

CHN1
Genomic Map