siRNA Details
| Gene Symbol: PSMA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025096.1 | NP_001020267.1 | 7 | 1534 | ||
| NM_001025097.1 | NP_001020268.1 | 7 | 1420 | |||
| NM_001308188.1 | NP_001295117.1 | 8 | 1604 | |||
| NM_144662.2 | NP_653263.2 | 7 | 1552 | |||
| XM_011525818.1 | XP_011524120.1 | 8 | 1487 | |||
| GenBank mRNA | BC021713.1 | - | 8 | 1544 | 4826048 | |
| BC025389.1 | - | 7 | 1530 | |||
| BC025393.1 | - | 7 | 1408 | |||
| BC028371.1 | - | 7 | 1407 | 4828792 | ||
| BC028686.1 | - | 7 | 1407 | 4828931 | ||
| BC047355.1 | - | 7 | 1526 | |||

PSMA8
Genomic Map