siRNA Details
| Gene Symbol: SENP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166340.1 | NP_001159812.1 | 2 | 1246 | ||
| NM_001172109.1 | NP_001165580.1 | 3 | 1400 | |||
| NM_001172110.1 | NP_001165581.1 | 3 | 1242 | |||
| NM_001172111.1 | NP_001165582.1 | 3 | 1259 | |||
| NM_145204.3 | NP_660205.3 | 2 | 1135 | |||
| XM_005254157.3 | XP_005254214.1 | 2 | 1015 | |||
| XM_011521215.2 | XP_011519517.1 | 3 | 1533 | |||
| XM_011521217.2 | XP_011519519.1 | 2 | 1123 | |||
| GenBank mRNA | AF308450.1 | - | 2 | 1039 | ||
| AK097557.1 | - | 2 | 1246 | |||
| AK292298.1 | - | 2 | 1137 | |||
| AY008293.1 | - | 2 | 1099 | |||
| BC031411.1 | - | 2 | 1012 | 5195120 | ||

SENP8
Genomic Map