siRNA Details
| Gene Symbol: KCND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004979.5 | NP_004970.3 | 1 | 1791 | ||
| XM_011543910.2 | XP_011542212.1 | 2 | 699 | |||
| XM_017029508.1 | XP_016884997.1 | 2 | 530 | |||
| XM_017029509.1 | XP_016884998.1 | 2 | 527 | |||
| XM_017029510.1 | XP_016884999.1 | 1 | 1641 | |||
| GenBank mRNA | AB021865.1 | - | 1 | 1516 | ||
| AF166003.1 | - | 1 | 268 | |||
| AJ005898.1 | - | 1 | 218 | |||
| AK315092.1 | - | 1 | 702 | |||
| BC045659.1 | - | 1 | 1789 | 5271042 | ||

KCND1
Genomic Map