siRNA Details
| Gene Symbol: NUDT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001248011.1 | NP_001234940.1 | 2 | 591 | ||
| NM_024047.4 | NP_076952.1 | 2 | 591 | |||
| NM_198038.2 | NP_932155.1 | 2 | 233 | |||
| XM_011532031.2 | XP_011530333.1 | 2 | 594 | |||
| XM_011532032.2 | XP_011530334.1 | 2 | 2770 | |||
| GenBank mRNA | AF265341.1 | - | 2 | 232 | ||
| AF273028.1 | - | 2 | 555 | |||
| AK027295.1 | - | 2 | 581 | |||
| AK074845.1 | - | 2 | 233 | |||
| AK075408.1 | - | 2 | 511 | |||
| AY026252.1 | - | 2 | 586 | |||
| AY359123.1 | - | 2 | 285 | |||
| BC000542.2 | - | 2 | 351 | 3163944 | ||
| BI552154.1 | - | 231 | ||||
| BM544951.1 | - | 2 | 603 | |||

NUDT9
Genomic Map