siRNA Details
| Gene Symbol: REN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000537.3 | NP_000528.1 | 2 | 163 | ||
| GenBank mRNA | AF117822.1 | - | 2 | 92 | ||
| BC033474.1 | - | 2 | 145 | 5188566 | ||
| BC047752.1 | - | 2 | 148 | 5590211 | ||
| BP321283.1 | - | 2 | 163 | |||
| CR536498.1 | - | 2 | 119 | |||

REN
Genomic Map