siRNA Details
| Gene Symbol: REN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000537.3 | NP_000528.1 | 2 | 186 | ||
| GenBank mRNA | AF117822.1 | - | 2 | 115 | ||
| BC033474.1 | - | 2 | 168 | 5188566 | ||
| BC047752.1 | - | 2 | 171 | 5590211 | ||
| BP321283.1 | - | 2 | 186 | |||
| CR536498.1 | - | 2 | 142 | |||

REN
Genomic Map