siRNA Details
| Gene Symbol: REN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000537.3 | NP_000528.1 | 9 | 1216 | ||
| GenBank mRNA | BC033474.1 | - | 9 | 1198 | 5188566 | |
| BC047752.1 | - | 9 | 1201 | 5590211 | ||
| CR536498.1 | - | 9 | 1172 | |||
| X00063.1 | - | 7 | 854 | |||

REN
Genomic Map