siRNA Details
| Gene Symbol: CTSV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201575.1 | NP_001188504.1 | 2, 3 | 168 | ||
| NM_001333.3 | NP_001324.2 | 2, 3 | 371 | |||
| GenBank mRNA | AB001928.1 | - | 2, 3 | 194 | ||
| AF070448.1 | - | 1, 2 | 148 | |||
| AK312806.1 | - | 2, 3 | 168 | |||
| AY358641.1 | - | 2, 3 | 371 | |||
| BC023504.2 | - | 2, 3 | 185 | 3139314 | ||
| BC110512.1 | - | 2, 3 | 240 | 40031744 | ||
| Y14734.1 | - | 2, 3 | 190 | |||

CTSV
Genomic Map