siRNA Details
| Gene Symbol: CTSG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001911.2 | NP_001902.1 | 4 | 401 | ||
| XM_011536499.1 | XP_011534801.1 | 4 | 421 | |||
| GenBank mRNA | AK225914.1 | - | 3 | 848 | ||
| BC014460.1 | - | 4 | 401 | 4859911 | ||
| CR456807.1 | - | 4 | 364 | |||
| CR541704.1 | - | 4 | 364 | |||
| M16117.1 | - | 4 | 372 | |||

CTSG
Genomic Map