siRNA Details
| Gene Symbol: PSMB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002795.3 | NP_002786.2 | 3, 4 | 413 | ||
| GenBank mRNA | BC013008.2 | - | 3, 4 | 376 | 3010175 | |
| D26598.1 | - | 3, 4 | 316 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PSMB3| Gene Symbol: PSMB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002795.3 | NP_002786.2 | 3, 4 | 413 | ||
| GenBank mRNA | BC013008.2 | - | 3, 4 | 376 | 3010175 | |
| D26598.1 | - | 3, 4 | 316 | |||