siRNA Details
| Gene Symbol: PSMB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002801.3 | NP_002792.1 | 8 | 904 | ||
| GenBank mRNA | AK225658.1 | - | 7 | 691 | ||
| AK312208.1 | - | 8 | 864 | |||
| BC017198.1 | - | 8 | 855 | 3143189 | ||
| BC052369.1 | - | 8 | 842 | 4054165 | ||
| BT019723.1 | - | 8 | 776 | |||
| BT019724.1 | - | 8 | 776 | |||
| CR456982.1 | - | 8 | 776 | |||
| Y13640.1 | - | 8 | 864 | |||

PSMB10
Genomic Map