siRNA Details
| Gene Symbol: CPZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014447.2 | NP_001014447.1 | 5 | 908 | ||
| NM_001014448.2 | NP_001014448.1 | 5 | 1171 | |||
| NM_003652.3 | NP_003643.2 | 4 | 875 | |||
| GenBank mRNA | AK056317.1 | - | 5 | 908 | ||
| AK127844.1 | - | 2 | 2244 | |||
| BC006393.2 | - | 5 | 1075 | |||
| BC018913.2 | - | 5 | 1075 | |||
| BC080539.1 | - | 5 | 842 | 5020194 | ||
| BF993977.1 | - | 2 | 264 | |||
| BX338482.2 | - | 4 | 729 | |||
| CV026874.1 | - | 3 | 323 | |||
| U83411.1 | - | 4 | 740 | |||

CPZ
Genomic Map