siRNA Details
| Gene Symbol: CPT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031847.2 | NP_001027017.1 | 5 | 653 | ||
| NM_001876.3 | NP_001867.2 | 5 | 653 | |||
| XM_005273762.2 | XP_005273819.1 | 5 | 629 | |||
| XM_005273763.1 | XP_005273820.1 | 5 | 629 | |||
| XM_017017220.1 | XP_016872709.1 | 5 | 762 | |||
| GenBank mRNA | AK172798.1 | - | 5 | 639 | ||
| AK309464.1 | - | 5 | 629 | |||
| AK314301.1 | - | 5 | 592 | |||
| BC000185.1 | - | 5 | 528 | 3352642 | ||
| BT009791.1 | - | 4 | 483 | |||
| L39211.1 | - | 5 | 559 | |||

CPT1A
Genomic Map