siRNA Details
| Gene Symbol: CST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001899.2 | NP_001890.1 | 1 | 139 | ||
| GenBank mRNA | BC065714.1 | - | 1 | 156 | 30327736 | |
| BC074952.2 | - | 1 | 112 | 30915278 | ||
| BC074953.2 | - | 1 | 112 | 30915500 | ||
| X54667.1 | - | 1 | 139 | |||

CST4
Genomic Map