siRNA Details
| Gene Symbol: TMPRSS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289000.1 | NP_001275929.1 | 3 | 315 | ||
| NM_001289001.1 | NP_001275930.1 | 3 | 325 | |||
| NM_153609.3 | NP_705837.1 | 3 | 394 | |||
| XM_011529989.2 | XP_011528291.1 | 3 | 453 | |||
| GenBank mRNA | AJ319876.1 | - | 2 | 257 | ||
| AY055384.1 | - | 3 | 394 | |||
| AY358398.1 | - | 3 | 325 | |||
| BC039082.1 | - | 3 | 397 | 5164881 | ||
| CR456446.1 | - | 3 | 315 | |||

TMPRSS6
Genomic Map