siRNA Details
| Gene Symbol: USP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182488.3 | NP_872294.2 | 3 | 464 | ||
| XM_005266282.2 | XP_005266339.1 | 3 | 378 | |||
| XM_011534956.2 | XP_011533258.1 | 2 | 132 | |||
| GenBank mRNA | AF022789.1 | - | 3 | 314 | ||
| AK289685.1 | - | 3 | 378 | |||
| BC026072.1 | - | 3 | 465 | 4830251 | ||

USP12
Genomic Map