siRNA Details
Gene Symbol: PRSS42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182702.1 | NP_874361.1 | 3 | 394 | ||
GenBank mRNA | AJ544583.1 | - | 3 | 394 | ||
AK092703.1 | - | 2 | 138 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PRSS42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182702.1 | NP_874361.1 | 3 | 394 | ||
GenBank mRNA | AJ544583.1 | - | 3 | 394 | ||
AK092703.1 | - | 2 | 138 |