siRNA Details
| Gene Symbol: PPP2R5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006243.3 | NP_006234.1 | 1, 2 | 763 | ||
| GenBank mRNA | AK312530.1 | - | 2, 3 | 578 | ||
| BC022474.1 | - | 1, 2 | 349 | 4815091 | ||
| BC110883.1 | - | 1, 2 | 472 | 6045358 | ||
| L42373.1 | - | 1, 2 | 760 | |||

PPP2R5A
Genomic Map