siRNA Details
| Gene Symbol: CTSE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317331.1 | NP_001304260.1 | 3 | 363 | ||
| NM_001910.3 | NP_001901.1 | 4 | 520 | |||
| NM_148964.2 | NP_683865.1 | 4 | 520 | |||
| XM_011509244.1 | XP_011507546.1 | 4 | 510 | |||
| XM_011509245.1 | XP_011507547.1 | 4 | 510 | |||
| GenBank mRNA | AJ250716.1 | - | 4 | 506 | ||
| AJ250717.1 | - | 4 | 506 | |||
| AK292057.1 | - | 4 | 450 | |||
| AK298069.1 | - | 3 | 216 | |||
| BC042537.1 | - | 4 | 487 | 5174814 | ||
| J05036.1 | - | 4 | 451 | |||

CTSE
Genomic Map