siRNA Details
| Gene Symbol: DUSP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007026.3 | NP_008957.1 | 3 | 488 | ||
| XM_005256977.3 | XP_005257034.1 | 3 | 1582 | |||
| XM_011524234.1 | XP_011522536.1 | 3 | 769 | |||
| GenBank mRNA | AF038844.1 | - | 3 | 427 | ||
| AF120032.1 | - | 1 | 196 | |||
| AK027210.1 | - | 2 | 1173 | |||
| AK314894.1 | - | 3 | 437 | |||
| BC000370.2 | - | 3 | 399 | 2819474 | ||
| BC001894.1 | - | 3 | 429 | 3533971 | ||
| BC004448.1 | - | 3 | 399 | 2819474 | ||
| CR536500.1 | - | 1 | 194 | |||
| CR541800.1 | - | 1 | 194 | |||

DUSP14
Genomic Map