siRNA Details
| Gene Symbol: DUSP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007240.2 | NP_009171.1 | 1 | 135 | ||
| XM_005244862.2 | XP_005244919.1 | 1 | 211 | |||
| GenBank mRNA | AF119226.1 | - | 1 | 112 | ||
| BC006286.1 | - | 1 | 133 | 3958403 | ||
| BT006633.1 | - | 1 | 101 | |||
| ES309317.1 | - | 1 | 151 | |||
| HY128542.1 | - | 1 | 136 | |||

DUSP12
Genomic Map