siRNA Details
| Gene Symbol: ILKAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030768.2 | NP_110395.1 | 3, 4 | 305 | ||
| XM_005246106.1 | XP_005246163.1 | 3, 4 | 298 | |||
| XM_006712784.1 | XP_006712847.1 | 3, 4 | 340 | |||
| XM_011511946.2 | XP_011510248.1 | 3, 4 | 302 | |||
| XM_017005056.1 | XP_016860545.1 | 3, 4 | 174 | |||
| XM_017005057.1 | XP_016860546.1 | 3, 4 | 688 | |||
| XM_017005058.1 | XP_016860547.1 | 3, 4 | 340 | |||
| GenBank mRNA | AK001043.1 | - | 3, 4 | 317 | ||
| AL136850.1 | - | 3, 4 | 230 | |||
| AY024365.1 | - | 3, 4 | 276 | |||
| BC006576.2 | - | 3, 4 | 305 | 3604852 | ||
| BC035746.1 | - | 2, 3 | 710 | |||
| CR533533.1 | - | 3, 4 | 175 | |||
| FJ491380.1 | - | 3, 4 | 198 | |||
| FJ491381.1 | - | 3, 4 | 175 | |||
| HQ658059.1 | - | 3, 4 | 305 | |||

ILKAP
Genomic Map