siRNA Details
| Gene Symbol: CFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317335.1 | NP_001304264.1 | 5 | 844 | ||
| NM_001928.3 | NP_001919.2 | 5 | 823 | |||
| GenBank mRNA | AK300963.1 | - | 5 | 818 | ||
| BC034529.1 | - | 4 | 739 | |||
| BC040146.1 | - | 4 | 725 | |||
| BC051001.1 | - | 5 | 763 | |||
| BC057807.1 | - | 5 | 822 | 5269709 | ||
| BI763909.1 | - | 199 | ||||
| M84526.1 | - | 5 | 771 | |||

CFD
Genomic Map