siRNA Details
| Gene Symbol: NCF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000433.3 | NP_000424.2 | 12 | 1348 | ||
| NM_001127651.2 | NP_001121123.1 | 13 | 1185 | |||
| NM_001190789.1 | NP_001177718.1 | 10 | 1105 | |||
| NM_001190794.1 | NP_001177723.1 | 11 | 1213 | |||
| XM_005245207.1 | XP_005245264.1 | 11 | 1169 | |||
| XM_011509580.1 | XP_011507882.1 | 13 | 1186 | |||
| XM_011509581.1 | XP_011507883.1 | 13 | 1173 | |||
| GenBank mRNA | AB209647.1 | - | 10 | 4618 | ||
| AF527950.1 | - | 12 | 1095 | |||
| AK296672.1 | - | 10 | 961 | |||
| AK298713.1 | - | 11 | 1122 | |||
| AK312666.1 | - | 12 | 1258 | |||
| BC001606.1 | - | 12 | 1325 | 3542841 | ||
| BT007439.1 | - | 12 | 1073 | |||
| KU178216.1 | - | 12 | 1073 | |||
| M32011.1 | - | 13 | 1140 | |||

NCF2
Genomic Map