siRNA Details
| Gene Symbol: CYP1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000499.4 | NP_000490.1 | 7 | 2219 | ||
| NM_001319216.1 | NP_001306145.1 | 6 | 2135 | |||
| NM_001319217.1 | NP_001306146.1 | 7 | 2222 | |||
| XM_017021953.1 | XP_016877442.1 | 8 | 2138 | |||
| GenBank mRNA | BC023019.1 | - | 7 | 2104 | 5123393 | |
| K03191.1 | - | 7 | 2109 | |||

CYP1A1
Genomic Map