siRNA Details
| Gene Symbol: RFXAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000538.3 | NP_000529.1 | 3 | 984 | ||
| GenBank mRNA | BC026088.1 | - | 3 | 963 | 4827378 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RFXAP| Gene Symbol: RFXAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000538.3 | NP_000529.1 | 3 | 984 | ||
| GenBank mRNA | BC026088.1 | - | 3 | 963 | 4827378 | |