siRNA Details
| Gene Symbol: ADH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000672.3 | NP_000663.1 | 8 | 1333 | ||
| GenBank mRNA | BC039065.1 | - | 7 | 1186 | 5184104 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADH6| Gene Symbol: ADH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000672.3 | NP_000663.1 | 8 | 1333 | ||
| GenBank mRNA | BC039065.1 | - | 7 | 1186 | 5184104 | |