siRNA Details
| Gene Symbol: CSNK1G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022048.3 | NP_071331.2 | 4, 5 | 780 | ||
| GenBank mRNA | AB042562.1 | - | 3, 4 | 428 | ||
| AB042563.1 | - | 3, 4 | 412 | |||
| AF223357.2 | - | 3, 4 | 498 | |||
| AK225960.1 | - | 4, 5 | 710 | |||
| AL832551.1 | - | 4, 5 | 558 | |||
| BC017236.2 | - | 3, 4 | 229 | |||
| BC130613.1 | - | 4, 5 | 553 | 40146644 | ||
| BC136504.1 | - | 4, 5 | 553 | 9020493 | ||

CSNK1G1
Genomic Map